Anti-HIST1H2AB Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:1000 to 1:2000
Type:
Antigen: HIST1H2AB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-983
- 77739-774
- Antigen Symbol:
- HIST1H2AB
- HIST1H2AB
- Antigen Synonyms:
- H2a 615|H2A|H2A GL101|H2A histone family member A|H2A.1|H2A.2|H2A/a|H2A/m|H2A/O|H2A/q|H2A1B_HUMAN|H2AFA|H2AFE|H2AFL|H2AFM|H2AFO|H2AFQ|HIST1H2AE|HIST1H2AJ|HIST2H2AA|HIST2H2AA3|HIST2H2AB|HIST2H2AC|Histone 1 H2ae|Histone 2 H2aa3|Histone 2 H2ab|Histone 2 H2ac|Histone H2A type 1 B|Histone H2A type 1 C|Histone H2A type 1 E|Histone H2A type 1 J|Histone H2A type 1-B/E|Histone H2A.2|Histone H2A/a|Histone H2A/m|MGC74460
- H2a 615|H2A|H2A GL101|H2A histone family member A|H2A.1|H2A.2|H2A/a|H2A/m|H2A/O|H2A/q|H2A1B_HUMAN|H2AFA|H2AFE|H2AFL|H2AFM|H2AFO|H2AFQ|HIST1H2AE|HIST1H2AJ|HIST2H2AA|HIST2H2AA3|HIST2H2AB|HIST2H2AC|Histone 1 H2ae|Histone 2 H2aa3|Histone 2 H2ab|Histone 2 H2ac|Histone H2A type 1 B|Histone H2A type 1 C|Histone H2A type 1 E|Histone H2A type 1 J|Histone H2A type 1-B/E|Histone H2A.2|Histone H2A/a|Histone H2A/m|MGC74460
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P04908
- P04908
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Histone H2A type 1-B/E protein (2-121AA)
- Recombinant Human Histone H2A type 1-B/E protein (2-121AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice