Anti-NDUFC2 Rabbit Polyclonal Antibody
77744-207
New Product
- Antigen Symbol:NDUFC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O95298
- Antigen Synonyms:B14.5b|CI-B14.5b|Complex I B14.5b|Complex I subunit B14.5b|Complex I-B14.5b|HLC-1|HLC1|Human lung cancer oncogene 1 protein|NADH dehydrogenase (ubiquinone) 1|subcomplex unknown|2|14.5kDa|NADH dehydrogenase [ubiquinone] 1 subunit C2|NADH dehydrogenase ubiquinone 1 subunit C2|NADH ubiquinone oxidoreductase subunit B14.5b|NADH-ubiquinone oxidoreductase subunit B14.5b|NADHDH2|NDUC2_HUMAN|NDUFC2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:B14.5b
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: NDUFC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human