Anti-NR2F6 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NR2F6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-961
- 77744-960
- Antigen Symbol:
- NR2F6
- NR2F6
- Antigen Synonyms:
- EAR 2|EAR-2|EAR2|ERBA RELATED 2|ERBA related gene 2|ERBAL2|NR2F6_HUMAN|Nuclear receptor subfamily 2 group F member 6|Orphan nuclear receptor EAR2 (V erbA related protein EAR 2)|v erb a avian erythroblastic leukemia viral oncogene homolog like|V erbA related protein EAR 2|V-erbA-related protein 2
- EAR 2|EAR-2|EAR2|ERBA RELATED 2|ERBA related gene 2|ERBAL2|NR2F6_HUMAN|Nuclear receptor subfamily 2 group F member 6|Orphan nuclear receptor EAR2 (V erbA related protein EAR 2)|v erb a avian erythroblastic leukemia viral oncogene homolog like|V erbA related protein EAR 2|V-erbA-related protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P10588
- P10588
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Nuclear receptor subfamily 2 group F member 6 protein (6-194AA)
- Nuclear receptor subfamily 2 group F member 6 protein (6-194AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice