Anti-ERC1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ERC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-094
- 77854-879
- Antigen Symbol:
- ERC1
- ERC1
- Antigen Synonyms:
- Cast 2|CAST2|CAZ associated structural protein 2|Elks|ELKS/RAB6 interacting/CAST family member 1|ELKS/RAB6-interacting/CAST family member 1|ERC 1|ERC protein 1|ERC-1|Erc1|ERC1 protein|KIAA1081|MGC12974|RAB6 interacting protein 2|RAB6-interacting protein 2|RAB6IP2|RB6I2_HUMAN
- Cast 2|CAST2|CAZ associated structural protein 2|Elks|ELKS/RAB6 interacting/CAST family member 1|ELKS/RAB6-interacting/CAST family member 1|ERC 1|ERC protein 1|ERC-1|Erc1|ERC1 protein|KIAA1081|MGC12974|RAB6 interacting protein 2|RAB6-interacting protein 2|RAB6IP2|RB6I2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IUD2
- Q8IUD2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human ELKS/Rab6-interacting/CAST family member 1 protein (1-224AA)
- Recombinant Human ELKS/Rab6-interacting/CAST family member 1 protein (1-224AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice