Anti-GFRA1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: GFRA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77738-175
- 77855-931
- Antigen Symbol:
- GFRA1
- GFRA1
- Antigen Synonyms:
- GDNF family receptor alpha 1|GDNF family receptor alpha-1|GDNF R|GDNF RA|GDNF receptor alpha|GDNF receptor alpha-1|GDNFR alpha 1|GDNFR alpha|GDNFR|GDNFR-alpha-1|GDNFRA|GDNFRalpha|GFR alpha 1|GFR alpha1|GFR-alpha-1|GFRA 1|GFRA1_HUMAN|GFRalpha1|Glial cell line derived neurotrophic factor receptor alpha|GPI linked anchor protein|MGC23045|PI linked cell surface accessory protein|RET 1L|RET ligand 1|RET1L|RETL 1|RETL1|TGF beta related neurotrophic factor receptor 1|TGF-beta-related neurotrophic factor receptor 1|TRNR 1|TRNR1
- GDNF family receptor alpha 1|GDNF family receptor alpha-1|GDNF R|GDNF RA|GDNF receptor alpha|GDNF receptor alpha-1|GDNFR alpha 1|GDNFR alpha|GDNFR|GDNFR-alpha-1|GDNFRA|GDNFRalpha|GFR alpha 1|GFR alpha1|GFR-alpha-1|GFRA 1|GFRA1_HUMAN|GFRalpha1|Glial cell line derived neurotrophic factor receptor alpha|GPI linked anchor protein|MGC23045|PI linked cell surface accessory protein|RET 1L|RET ligand 1|RET1L|RETL 1|RETL1|TGF beta related neurotrophic factor receptor 1|TGF-beta-related neurotrophic factor receptor 1|TRNR 1|TRNR1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P56159
- P56159
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human GDNF family receptor alpha-1 protein (230-385AA)
- Recombinant human GDNF family receptor alpha-1 protein (230-385AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice