Anti-GLI3 Rabbit Polyclonal Antibody
Catalog # 77737-522
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GLI3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:P10071
- Antigen Synonyms:ACLS|DNA binding protein|GCPS|Gli 3|GLI family zinc finger 3|GLI Kruppel family member GLI 3|GLI Kruppel family member GLI3 (Greig cephalopolysyndactyly syndrome)|GLI Kruppel family member GLI3 C-terminally truncated form|GLI3 form of 190 kDa|GLI3 form of 83 kDa|GLI3 full length protein|GLI3-190|GLI3-83|GLI3_HUMAN|GLI3FL|Glioma associated oncogene family zinc finger 3|Oncogene PAP A|PAPA 1|PAPA|PAPA1|PAPB|PHS|PPD IV|PPDIV|Transcriptional activator Transcriptional repressor GLI3R|Zinc finger protein GLI 3|Zinc finger protein GLI3
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:ACLS
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA:1:10000
Type:
Antigen: GLI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat