Anti-KCNA2 Rabbit Polyclonal Antibody
Catalog # 77741-440
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:KCNA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P16389
- Antigen Synonyms:KCNA2Potassium voltage-gated channel subfamily A member 2|NGK1|Voltage-gated K(+) channel HuKIV|Voltage-gated potassium channel HBK5|Voltage-gated potassium channel subunit Kv1.2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Voltage-gated potassium channel subunit Kv1.2
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: KCNA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat