Anti-GLYCTK Rabbit Polyclonal Antibody
Catalog # 77738-480
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GLYCTK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8IVS8
- Antigen Synonyms:CG9886 like|GLCTK_HUMAN|Glycerate kinase|Glyctk|GLYCTK1|HBeAg binding protein 2|HBeAg binding protein 4|HBeAg-binding protein 4|HBeAgBP4A|HBEBP2|HBEBP4
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:HBEBP4
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: GLYCTK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat