Anti-GAS1 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
0
Type:
Antigen: GAS1
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77737-985
- 77855-807
- Antigen Symbol:
- GAS1
- GAS1
- Antigen Synonyms:
- GAS-1| GAS1_HUMAN| Growth arrest-specific protein 1| Growth-arrest-specific protein 1 precursor
- GAS-1|GAS1_HUMAN|Growth arrest-specific protein 1|Growth-arrest-specific protein 1 precursor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P54826
- P54826
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Growth arrest-specific protein 1 protein (248-318AA)
- Recombinant Human Growth arrest-specific protein 1 protein (248-318AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice