Anti-NCAPH Rabbit Polyclonal Antibody
77743-984
New Product
- Antigen Symbol:NCAPH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q15003
- Antigen Synonyms:Barren Drosophila Homolog of 1| Barren homolog protein 1| BRRN| BRRN1| CAP H| CAPH| Chromosome associated protein H| Chromosome-associated protein H| CND2_HUMAN| Condensin complex subunit 2| Condensin subunit 2| Condensin-2 complex subunit H2| Condesin| HCAP H| hCAP-H| KIAA0074| mCAP H| MGC4616| Non SMC condensin I complex subunit H| Non-SMC condensin I complex subunit H| XCAP H homolog| XCAP-H homolog
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:XCAP-H homolog
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: NCAPH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat