Anti-KCNJ3 Rabbit Polyclonal Antibody
Catalog # 77740-115
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:KCNJ3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:P48549
- Antigen Synonyms:G protein activated inward rectifier potassium channel 1|G protein-activated inward rectifier potassium channel 1|GIRK-1|GIRK1|Inward rectifier K(+) channel Kir3.1|Inwardly rectifying potassium channel KIR3.1|inwardly rectifying subfamily J member 3|IRK3|IRK3_HUMAN|Kcnf3|Kcnj3|KGA|KGB1|Kir3.1|Potassium channel|Potassium inwardly-rectifying channel|subfamily J|member 3
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:G protein activated inward rectifier potassium channel 1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:2000, IF:1:200 to 1:1000, ELISA:1:20000
Type:
Antigen: KCNJ3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat