Anti-MAP2K4 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: MAP2K4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77742-641
- 77859-370
- Antigen Symbol:
- MAP2K4
- Antigen Synonyms:
- c Jun N terminal kinase kinase 1|C-JUN N-terminal kinase kinase 1|Dual specificity mitogen activated protein kinase kinase 4|Dual specificity mitogen-activated protein kinase kinase 4|JNK Activated Kinase 1|JNK activating kinase 1|JNK-activating kinase 1|JNKK|JNKK1|MAP kinase kinase 4|Map2k4|MAPK ERK kinase 4|MAPK/ERK kinase 4|MAPKK 4|MAPKK4|MEK 4|MEK4|Mitogen activated protein kinase kinase 4|MKK 4|MKK4|MP2K4_HUMAN|PRKMK4|SAPK ERK kinase 1|SAPK/ERK kinase 1|SAPKK 1|SAPKK1|SEK1|SERK1|SKK1|Stress activated protein kinase kinase 1
- c Jun N terminal kinase kinase 1|C-JUN N-terminal kinase kinase 1|Dual specificity mitogen activated protein kinase kinase 4|Dual specificity mitogen-activated protein kinase kinase 4|JNK Activated Kinase 1|JNK activating kinase 1|JNK-activating kinase 1|JNKK|JNKK1|MAP kinase kinase 4|Map2k4|MAPK ERK kinase 4|MAPK/ERK kinase 4|MAPKK 4|MAPKK4|MEK 4|MEK4|Mitogen activated protein kinase kinase 4|MKK 4|MKK4|MP2K4_HUMAN|PRKMK4|SAPK ERK kinase 1|SAPK/ERK kinase 1|SAPKK 1|SAPKK1|SEK1|SERK1|SKK1|Stress activated protein kinase kinase 1
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P45985
- P45985
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Dual specificity mitogen-activated protein kinase kinase 4 protein (259-398AA)
- Recombinant Human Dual specificity mitogen-activated protein kinase kinase 4 protein (259-398AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice