Anti-L2HGDH Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: L2HGDH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Specifications
- Catalog No:
- 77858-800
- 77741-936
- Antigen Symbol:
- L2HGDH
- Antigen Synonyms:
- L2HGDHL-2-hydroxyglutarate dehydrogenase|mitochondrial|EC 1.1.99.2|Duranin
- L2HGDHL-2-hydroxyglutarate dehydrogenase|mitochondrial|EC 1.1.99.2|Duranin
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Reactivity:
- Bovine
- Host:
- Rabbit
- Gene ID:
- A7MBI3
- A7MBI3
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Bovine L-2-hydroxyglutarate dehydrogenase, mitochondrial protein (53-463AA)
- Recombinant Bovine L-2-hydroxyglutarate dehydrogenase, mitochondrial protein (53-463AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice