Anti-MRPL42 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MRPL42
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-422
- 77859-967
- Antigen Symbol:
- MRPL42
- MRPL42
- Antigen Synonyms:
- 28S ribosomal protein S32|39S ribosomal protein L31|39S ribosomal protein L42|HSPC204|L31mt|L42mt|mitochondrial|Mitochondrial ribosomal protein L42|Mitochondrial ribosomal protein S32|MRP L31|MRP L42|MRP S32|MRP-L31|MRP-L42|MRP-S32|MRPL31|MRPS32|PTD007|RM42_HUMAN|RPML31|S32mt
- 28S ribosomal protein S32|39S ribosomal protein L31|39S ribosomal protein L42|HSPC204|L31mt|L42mt|mitochondrial|Mitochondrial ribosomal protein L42|Mitochondrial ribosomal protein S32|MRP L31|MRP L42|MRP S32|MRP-L31|MRP-L42|MRP-S32|MRPL31|MRPS32|PTD007|RM42_HUMAN|RPML31|S32mt
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6G3
- Q9Y6G3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human 39S ribosomal protein L42, mitochondrial protein (33-142AA)
- Recombinant Human 39S ribosomal protein L42, mitochondrial protein (33-142AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice