Anti-ENPP6 Rabbit Polyclonal Antibody
Catalog # 77735-933
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ENPP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q6UWR7
- Antigen Synonyms:UNQ1889/PRO4334Glycerophosphocholine cholinephosphodiesterase GPC-Cpde|EC 3.1.4.-|EC 3.1.4.38|Choline-specific glycerophosphodiester phosphodiesterase|Ectonucleotide pyrophosphatase/phosphodiesterase family member 6|E-NPP 6|NPP-6
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:NPP-6
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ENPP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat