Anti-HIST1H2BC Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:50 to 1:500, IHC: 1:10 to 1:100, IF: 1:1 to 1:10
Type:
Antigen: HIST1H2BC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Specifications
- Catalog No:
- 77739-853
- 77857-116
- Antigen Symbol:
- HIST1H2BC
- HIST1H2BC
- Antigen Synonyms:
- H2BC4|H2BFL|H2BC6|H2BFH|HIST1H2BE|H2BC7|H2BFG|HIST1H2BF|H2BC8|H2BFA|HIST1H2BG|H2BC10|H2BFK|HIST1H2BIHistone H2B type 1-C/E/F/G/I|Histone H2B.1 A|Histone H2B.a|H2B/a|Histone H2B.g|H2B/g|Histone H2B.h|H2B/h|Histone H2B.k|H2B/k|Histone H2B.l|H2B/l
- H2BC4|H2BFL|H2BC6|H2BFH|HIST1H2BE|H2BC7|H2BFG|HIST1H2BF|H2BC8|H2BFA|HIST1H2BG|H2BC10|H2BFK|HIST1H2BIHistone H2B type 1-C/E/F/G/I|Histone H2B.1 A|Histone H2B.a|H2B/a|Histone H2B.g|H2B/g|Histone H2B.h|H2B/h|Histone H2B.k|H2B/k|Histone H2B.l|H2B/l
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P62807
- P62807
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Peptide sequence around site of Lys (20) derived from Human Histone H2B type 1-C/E/F/G/I
- Peptide sequence around site of Lys (20) derived from Human Histone H2B type 1-C/E/F/G/I
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice