Anti-CXCL3 Rabbit Polyclonal Antibody
77737-680
New Product
- Antigen Symbol:CXCL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P19876
- Antigen Synonyms:C-X-C motif chemokine 3|C-X-C motif chemokine ligand 3|Chemokine (C X C motif) ligand 3|Chemokine (CXC motif) ligand 3|Cinc 2|CINC 2b|Cinc2|CINC2b|CXCL 3|CXCL3_HUMAN|Cytokine induced neutrophil chemoattractant 2|Dcip1|Dendritic cell inflammatory protein 1|Gm1960|GRO protein gamma|GRO-gamma|GRO-gamma(1-73)|GRO-gamma(5-73)|GRO3|GRO3 oncogene|GROG|Growth regulated protein gamma|Growth-regulated protein gamma|Macrophage inflammatory protein 2 beta precursor|Macrophage inflammatory protein 2-beta|Melanoma growth stimulatory activity gamma|Member 3|MGSA gamma|MIP 2b|MIP2-beta|MIP2B|SCYB3|Small inducible cytokine subfamily B
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:C-X-C motif chemokine 3
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:10000
Type:
Antigen: CXCL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human