Anti-GPT Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GPT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-936
- 77856-393
- Antigen Symbol:
- GPT
- GPT
- Antigen Synonyms:
- AAT1|Alanine aminotransferase 1|Alanine aminotransferase|ALAT1_HUMAN|ALT1|Glutamate pyruvate transaminase 1|Glutamic alanine transaminase 1|Glutamic pyruvate transaminase (alanine aminotransferase)|Glutamic pyruvic transaminase 1|Glutamic--alanine transaminase 1|Glutamic--pyruvic transaminase 1|GPT 1|GPT1
- AAT1|Alanine aminotransferase 1|Alanine aminotransferase|ALAT1_HUMAN|ALT1|Glutamate pyruvate transaminase 1|Glutamic alanine transaminase 1|Glutamic pyruvate transaminase (alanine aminotransferase)|Glutamic pyruvic transaminase 1|Glutamic--alanine transaminase 1|Glutamic--pyruvic transaminase 1|GPT 1|GPT1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P24298
- P24298
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Alanine aminotransferase 1 protein (2-496AA)
- Recombinant Human Alanine aminotransferase 1 protein (2-496AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice