Anti-KRT81 Rabbit Polyclonal Antibody
Catalog # 77741-906
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:KRT81
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q14533
- Antigen Synonyms:1|basic|ghHb 1|ghHb1|ghHkb 1|ghHKb1|hair|Hair keratin K2.9|Hard keratin type II 1|HB 1|HB1|hHAKB2 1|K2.9|K81|Keratin 81|Keratin|Keratin hair basic 1|Keratin type II cuticular Hb1|Keratin-81|Keratin81|KRT 81|KRT81_HUMAN|KRTHB 1|KRTHB1|Metastatic lymph node 137 gene protein|MLN 137|MLN137|type II cuticular Hb1|Type II hair keratin Hb1|Type-II keratin Kb21
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Type-II keratin Kb21
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: KRT81
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat