Anti-NOP2 Rabbit Polyclonal Antibody
Catalog # 77744-641
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NOP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P46087
- Antigen Synonyms:NOL1|NSUN1Probable 28S rRNA|cytosine(4447)-C(5))-methyltransferase|EC 2.1.1.-|Nucleolar protein 1|Nucleolar protein 2 homolog|Proliferating-cell nucleolar antigen p120|Proliferation-associated nucleolar protein p120
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Proliferation-associated nucleolar protein p120
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: NOP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human