Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-OGG1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-OGG1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
Anti-OGG1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77745-397
  • 77745-396
  • Antigen Symbol:
  • OGG1
  • OGG1
  • Antigen Synonyms:
  • 8 hydroxyguanine DNA glycosylase|8 oxoguanine DNA glycosylase 1|8 oxoguanine DNA glycosylase|AP lyase|DNA (apurinic or apyrimidinic site) lyase|DNA apurinic or apyrimidinic site lyase|DNA lyase|DNA-(apurinic or apyrimidinic site) lyase|HMMH|HOGG 1|HOGG1|MMH|MUTM|N glycosylase|Ogg 1|Ogg1|OGG1_HUMAN|OGH 1|OGH1
  • 8 hydroxyguanine DNA glycosylase|8 oxoguanine DNA glycosylase 1|8 oxoguanine DNA glycosylase|AP lyase|DNA (apurinic or apyrimidinic site) lyase|DNA apurinic or apyrimidinic site lyase|DNA lyase|DNA-(apurinic or apyrimidinic site) lyase|HMMH|HOGG 1|HOGG1|MMH|MUTM|N glycosylase|Ogg 1|Ogg1|OGG1_HUMAN|OGH 1|OGH1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O15527
  • O15527
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • N-glycosylase/DNA lyase protein (1-345AA)
  • N-glycosylase/DNA lyase protein (1-345AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95% Protein G purified
  • >95% Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications