Anti-GRIN2D Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: GRIN2D
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-040
- 77856-464
- Antigen Symbol:
- GRIN2D
- GRIN2D
- Antigen Synonyms:
- EB11|estrogen receptor binding CpG island|GluN2D|Glutamate [NMDA] receptor subunit epsilon 4|Glutamate [NMDA] receptor subunit epsilon-4|Glutamate receptor ionotropic|glutamate receptor ionotropic NMDA 2D|Glutamate receptor|ionotropic|N-methyl D-aspartate 2D|N methyl D aspartate receptor subtype 2D|N methyl d aspartate receptor subunit 2D|N-methyl D-aspartate receptor subtype 2D|NMDA 2D|NMDAR2D|NMDE4|NMDE4_HUMAN|NR2D
- EB11|estrogen receptor binding CpG island|GluN2D|Glutamate [NMDA] receptor subunit epsilon 4|Glutamate [NMDA] receptor subunit epsilon-4|Glutamate receptor ionotropic|glutamate receptor ionotropic NMDA 2D|Glutamate receptor|ionotropic|N-methyl D-aspartate 2D|N methyl D aspartate receptor subtype 2D|N methyl d aspartate receptor subunit 2D|N-methyl D-aspartate receptor subtype 2D|NMDA 2D|NMDAR2D|NMDE4|NMDE4_HUMAN|NR2D
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15399
- O15399
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Glutamate receptor ionotropic, NMDA 2D protein (330-480AA)
- Recombinant Human Glutamate receptor ionotropic, NMDA 2D protein (330-480AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice