Anti-MEFV Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC:1:200 to 1:500
Type:
Antigen: MEFV
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77859-643
- 77742-985
- Antigen Symbol:
- MEFV
- MEFV
- Antigen Synonyms:
- FMF|Marenostrin|Mediterranean fever|Mediterranean fever protein|MEF|MEFV_HUMAN|Pyrin|TRIM20
- FMF|Marenostrin|Mediterranean fever|Mediterranean fever protein|MEF|MEFV_HUMAN|Pyrin|TRIM20
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15553
- O15553
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Pyrin protein (268-374AA)
- Recombinant Human Pyrin protein (268-374AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice