Anti-NAT8 Rabbit Polyclonal Antibody
77743-960
New Product
- Antigen Symbol:NAT8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UHE5
- Antigen Synonyms:acetyltransferase 2|ATase2|Camello, Xenopus, homolog of, 1|Camello-like protein 1|CML1|cysteinyl-conjugate N-acetyltransferase|GLA|Hcml1|kidney- and liver-specific gene product|N-acetyltransferase 8 (camello like)|N-acetyltransferase 8 (GCN5-related, putative)|N-acetyltransferase 8|NAT8_HUMAN|Probable N-acetyltransferase 8|TSC501|TSC510
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:TSC510
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human