Anti-FDFT1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: FDFT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77737-015
- 77855-278
- Antigen Symbol:
- FDFT1
- FDFT1
- Antigen Synonyms:
- DGPT|ERG9|Farnesyl diphosphate farnesyltransferase|Farnesyl-diphosphate farnesyltransferase|FDFT_HUMAN|FPP:FPP farnesyltransferase|SQS|Squalene synthase|Squalene synthetase|SS
- DGPT|ERG9|Farnesyl diphosphate farnesyltransferase|Farnesyl-diphosphate farnesyltransferase|FDFT_HUMAN|FPP:FPP farnesyltransferase|SQS|Squalene synthase|Squalene synthetase|SS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P37268
- P37268
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Squalene synthase protein (2-417AA)
- Recombinant Human Squalene synthase protein (2-417AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice