Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-NDUFA6 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-NDUFA6 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
Anti-NDUFA6 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77744-149
  • 77744-148
  • Antigen Symbol:
  • NDUFA6
  • NDUFA6
  • Antigen Synonyms:
  • B14|CI-B14|complex I B14 subunit|Complex I-B14|LYR motif-containing protein 6|LYRM6|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6 (14kD, B14)|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6, 14kDa|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 6|NADH-ubiquinone oxidoreductase 1 alpha subcomplex, 6|NADH-ubiquinone oxidoreductase B14 subunit|NADHB14|NDUA6_HUMAN|NDUFA6
  • B14|CI-B14|complex I B14 subunit|Complex I-B14|LYR motif-containing protein 6|LYRM6|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6 (14kD, B14)|NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6, 14kDa|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 6|NADH-ubiquinone oxidoreductase 1 alpha subcomplex, 6|NADH-ubiquinone oxidoreductase B14 subunit|NADHB14|NDUA6_HUMAN|NDUFA6
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P56556
  • P56556
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 6 protein (27-154AA)
  • NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 6 protein (27-154AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications