Anti-MSH6 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: MSH6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77860-028
- 77743-515
- Antigen Symbol:
- MSH6
- MSH6
- Antigen Synonyms:
- DNA mismatch repair protein G/T mismatch binding protein|G/T mismatch-binding protein|GTBP|GTMBP|hHNPCC 5|HNPCC5|HSAP|MSH 6|MSH6_HUMAN|mutS (E. coli) homolog 6|MutS alpha 160 kDa subunit|MutS homolog 6 (E. coli)|mutS homolog 6|MutS-alpha 160 kDa subunit|p160|Sperm associated protein
- DNA mismatch repair protein G/T mismatch binding protein|G/T mismatch-binding protein|GTBP|GTMBP|hHNPCC 5|HNPCC5|HSAP|MSH 6|MSH6_HUMAN|mutS (E. coli) homolog 6|MutS alpha 160 kDa subunit|MutS homolog 6 (E. coli)|mutS homolog 6|MutS-alpha 160 kDa subunit|p160|Sperm associated protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52701
- P52701
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human DNA mismatch repair protein Msh6 protein (1-400AA)
- Recombinant Human DNA mismatch repair protein Msh6 protein (1-400AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice