Anti-NEK4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: NEK4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-304
- 77744-305
- Antigen Symbol:
- NEK4
- NEK4
- Antigen Synonyms:
- STK2|Serine/threonine-protein kinase EC 2.7.11.1|Never in mitosis A-related kinase 4|NimA-related protein kinase 4|Serine/threonine-protein kinase 2|Serine/threonine-protein kinase NRK2
- STK2|Serine/threonine-protein kinase EC 2.7.11.1|Never in mitosis A-related kinase 4|NimA-related protein kinase 4|Serine/threonine-protein kinase 2|Serine/threonine-protein kinase NRK2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P51957
- P51957
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Serine/threonine-protein kinase Nek4 protein (516-661AA)
- Serine/threonine-protein kinase Nek4 protein (516-661AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice