Anti-FKBP3 Rabbit Polyclonal Antibody
Catalog # 77737-236
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FKBP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q00688
- Antigen Synonyms:25 kDa FK506-binding protein|25 kDa FKBP|FK506 binding protein 25 T cell|FK506 binding protein 25|T cell|25-KD|FK506 binding protein 3 (25kD)|FK506-binding protein 3|FKBP 25|FKBP 3|FKBP-25|FKBP-3|FKBP25|FKBP3|FKBP3_HUMAN|Immunophilin FKBP25|Peptidyl-prolyl cis-trans isomerase FKBP3|PPIase|PPIase FKBP3|Rapamycin binding protein|Rapamycin-selective 25 kDa immunophilin|Rotamase
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Rotamase
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: FKBP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat