Anti-GUCA1A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: GUCA1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77739-267
- 77856-608
- Antigen Symbol:
- GUCA1A
- GUCA1A
- Antigen Synonyms:
- C6orf131|GCAP|GCAP1|GUCA1|Guanylyl cyclase-activating protein 1|GCAP 1|Guanylate cyclase activator 1A
- C6orf131|GCAP|GCAP1|GUCA1|Guanylyl cyclase-activating protein 1|GCAP 1|Guanylate cyclase activator 1A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P43080
- P43080
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Guanylyl cyclase-activating protein 1 protein (2-201AA)
- Recombinant Human Guanylyl cyclase-activating protein 1 protein (2-201AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice