Anti-PRAG1 Rabbit Polyclonal Antibody
Catalog # 77735-766
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:PRAG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q86YV5
- Antigen Synonyms:D8Ertd82e|DKFZp761P0423|DNA segment, Chr 8, ERATO Doi 82|EC 2.7.10.2|Homolog of rat pragma of Rnd2|LOC157285|PRAGMIN|SG223_HUMAN|Sgk223|Sugen kinase 223|Tyrosine protein kinase SgK223|Tyrosine-protein kinase SgK223
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:D8Ertd82e
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:10000
Type:
Antigen: PRAG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat