Anti-NOVA2 Rabbit Polyclonal Antibody
Catalog # 77744-706
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NOVA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UNW9
- Antigen Synonyms:NOVA3RNA-binding protein Nova-2|Astrocytic NOVA1-like RNA-binding protein|ANOVA|Neuro-oncological ventral antigen 2
- Storage Buffer:0.1 M NaHCO₃, 0.1 M glycine, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Neuro-oncological ventral antigen 2
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: NOVA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat