Anti-MED30 Rabbit Polyclonal Antibody
Catalog # 77742-965
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MED30
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96HR3
- Antigen Synonyms:MED30_HUMAN|Mediator complex subunit 30|Mediator of RNA polymerase II transcription subunit 30|THRAP6|Thyroid hormone receptor-associated protein 6|Thyroid hormone receptor-associated protein complex 25 kDa component|TRAP/Mediator complex component TRAP25|Trap25
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Trap25
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MED30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat