Anti-ID2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ID2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-569
- 77857-785
- Antigen Symbol:
- ID2
- ID2
- Antigen Synonyms:
- bHLHb26|Cell growth inhibiting gene 8|class B basic helix loop helix protein 26|Class B basic helix-loop-helix protein 26|DNA binding protein inhibitor ID 2|DNA binding protein inhibitor ID2|DNA-binding protein inhibitor ID-2|GIG 8|GIG8|Helix loop helix protein ID2|ID2|ID2_HUMAN|ID2A|ID2H|Inhibitor of differentiation 2|Inhibitor of DNA binding 2|Inhibitor of DNA binding 2, dominant negative helix loop helix protein|MGC26389
- Inhibitor of DNA binding 2|Cell growth inhibiting gene 8|class B basic helix loop helix protein 26|DNA binding protein inhibitor ID2|Inhibitor of DNA binding 2, dominant negative helix loop helix protein|DNA-binding protein inhibitor ID-2|ID2A|ID2_HUMAN|Helix loop helix protein ID2|bHLHb26|ID2H|MGC26389|Class B basic helix-loop-helix protein 26|GIG8|GIG 8|ID2|DNA binding protein inhibitor ID 2|Inhibitor of differentiation 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02363
- Q02363
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DNA-binding protein inhibitor ID-2 protein (1-134AA)
- Recombinant Human DNA-binding protein inhibitor ID-2 protein (1-134AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice