Anti-HIST1H2BC Rabbit Polyclonal Antibody
New Product
Recommended dilution: IHC:1:10-1:100, IF:1:1-1:10
Type:
Antigen: HIST1H2BC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77857-097
- 77739-844
- Antigen Symbol:
- HIST1H2BC
- HIST1H2BC
- Antigen Synonyms:
- H2BC4|H2BFL||H2BC6|H2BFH|HIST1H2BE|H2BC7|H2BFG|HIST1H2BF|H2BC8|H2BFA|HIST1H2BG|H2BC10|H2BFK|HIST1H2BIHistone H2B type 1-C/E/F/G/I|Histone H2B.1 A|Histone H2B.a|H2B/a|Histone H2B.g|H2B/g|Histone H2B.h|H2B/h|Histone H2B.k|H2B/k|Histone H2B.l|H2B/l
- H2BC4| H2BFL| | H2BC6| H2BFH| HIST1H2BE|| H2BC7| H2BFG| HIST1H2BF| H2BC8| H2BFA| HIST1H2BG| H2BC10| H2BFK| HIST1H2BIHistone H2B type 1-C/E/F/G/I| Histone H2B.1 A| Histone H2B.a| H2B/a| Histone H2B.g| H2B/g| Histone H2B.h| H2B/h| Histone H2B.k| H2B/k| Histone H2B.l| H2B/l
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P62807
- P62807
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Peptide sequence around site of Lys (12) derived from Human Histone H2B type 1-C/E/F/G/I
- Peptide sequence around site of Lys (12) derived from Human Histone H2B type 1-C/E/F/G/I
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice