Anti-GTF2E2 Rabbit Polyclonal Antibody
Catalog # 77739-215
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GTF2E2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P29084
- Antigen Synonyms:Transcription initiation factor IIE beta subunit|polypeptide 2|Transcription initiation factor IIE subunit beta|TF2E2|TFIIE B|beta 34kDa|T2EB_HUMAN|TFIIE-beta|General transcription factor IIE subunit 2|General transcription factor IIE|TF2E 2|FE
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Transcription initiation factor IIE subunit beta
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: GTF2E2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat