Anti-OR8H2 Rabbit Polyclonal Antibody
Catalog # 77738-779
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:OR8H2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q8N162
- Antigen Synonyms:Olfactory receptor 8H2|Olfactory receptor OR11-171
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Olfactory receptor OR11-171
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IF: 1:200 to 1:1000, ELISA: 1:20000
Type:
Antigen: OR8H2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human