Anti-GPN3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: GPN3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77856-325
- 77738-767
- Antigen Symbol:
- GPN3
- Antigen Synonyms:
- ATP-binding domain 1 family member C|ATPBD1C|GPN-loop GTPase 3|Gpn3|GPN3_HUMAN
- ATP-binding domain 1 family member C|ATPBD1C|GPN-loop GTPase 3|Gpn3|GPN3_HUMAN
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- Q9UHW5
- Q9UHW5
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human GPN-loop GTPase 3 protein (181-284AA)
- Recombinant Human GPN-loop GTPase 3 protein (181-284AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice