Anti-GAS7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GAS7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-997
- 77855-816
- Antigen Symbol:
- GAS7
- GAS7
- Antigen Synonyms:
- GAS 7|GAS-7|GAS7_HUMAN|Growth arrest specific 7|Growth Arrest Specific Protein 7|Growth arrest-specific protein 7|KIAA0394|MGC1348|MLL/MLL/GAS7 fusion protein
- GAS 7|GAS-7|GAS7_HUMAN|Growth arrest specific 7|Growth Arrest Specific Protein 7|Growth arrest-specific protein 7|KIAA0394|MGC1348|MLL/MLL/GAS7 fusion protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60861
- O60861
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Growth arrest-specific protein 7 protein (1-153AA)
- Recombinant human Growth arrest-specific protein 7 protein (1-153AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice