Anti-MPG Rabbit Polyclonal Antibody
Catalog # 77743-315
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MPG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P29372
- Antigen Synonyms:3 alkyladenine DNA glycosylase|3-alkyladenine DNA glycosylase|3-methyladenine DNA glycosidase|3MG_HUMAN|AAG|ADPG|Alkyladenine DNA glycosylase|anpg|APNG|CRA36.1|DNA 3 methyladenine glycosylase|DNA-3-methyladenine glycosylase|MDG|Mid1|Mpg|N methylpurine DNA glycosirase|N methylpurine DNA glycosylase|N-methylpurine-DNA glycosylase|PIG11|PIG16|Proliferation inducing protein 11|Proliferation inducing protein 16
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Proliferation inducing protein 16
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat