Anti-MTF2 Rabbit Polyclonal Antibody
Catalog # 77743-582
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MTF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9Y483
- Antigen Synonyms:PCL2|Metal-response element-binding transcription factor 2|Metal regulatory transcription factor 2|Metal-response element DNA-binding protein M96|Polycomb-like protein 2|hPCl2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:hPCl2
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human