Anti-GRB2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GRB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-427
- 77738-988
- Antigen Symbol:
- GRB2
- GRB2
- Antigen Synonyms:
- Abundant SRC homology|Adapter protein ASH|Ash protein|EGFRBP Epidermal growth factor receptor binding protein|Epidermal growth factor receptor binding protein GRB 2|GRB2 adapter protein|GRB2_HUMAN|Grb3 3|Growth factor receptor bound protein 2|Growth factor receptor bound protein 3|Growth factor receptor-bound protein 2|HT027|MST084|MSTP084|NCKAP2|OTTHUMP00000166096|OTTHUMP00000166097|OTTHUMP00000166098|Protein ash|SEM5|SH2/SH3 adapter GRB2
- Abundant SRC homology|Adapter protein ASH|Ash protein|EGFRBP Epidermal growth factor receptor binding protein|Epidermal growth factor receptor binding protein GRB 2|GRB2 adapter protein|GRB2_HUMAN|Grb3 3|Growth factor receptor bound protein 2|Growth factor receptor bound protein 3|Growth factor receptor-bound protein 2|HT027|MST084|MSTP084|NCKAP2|OTTHUMP00000166096|OTTHUMP00000166097|OTTHUMP00000166098|Protein ash|SEM5|SH2/SH3 adapter GRB2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P62993
- P62993
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Growth factor receptor-bound protein 2 protein (11-200AA)
- Recombinant Human Growth factor receptor-bound protein 2 protein (11-200AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice