Anti-GDI1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: GDI1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77858-554
- 77741-628
- Antigen Symbol:
- GDI1
- GDI1
- Antigen Synonyms:
- 1A|FLJ41411|GDI-1|gdi1|GDIA_HUMAN|GDIL|GDP dissociation inhibitor 1|Guanosine diphosphate dissociation inhibitor 1|Mental retardation X-linked 41|Mental retardation X-linked 48|MRX41|MRX48|Oligophrenin 2|Oligophrenin-2|OPHN2|Protein XAP 4|Protein XAP-4|Protein XAP4|Rab GDI alpha|Rab GDP dissociation inhibitor alpha|RABGD1A|RABGDIA|XAP 4
- 1A|FLJ41411|GDI-1|gdi1|GDIA_HUMAN|GDIL|GDP dissociation inhibitor 1|Guanosine diphosphate dissociation inhibitor 1|Mental retardation X-linked 41|Mental retardation X-linked 48|MRX41|MRX48|Oligophrenin 2|Oligophrenin-2|OPHN2|Protein XAP 4|Protein XAP-4|Protein XAP4|Rab GDI alpha|Rab GDP dissociation inhibitor alpha|RABGD1A|RABGDIA|XAP 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31150
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Rab GDP dissociation inhibitor alpha protein (35-170AA)
- Recombinant Human Rab GDP dissociation inhibitor alpha protein (35-170AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice