Anti-FKBP6 Rabbit Polyclonal Antibody
77737-248
New Product
- Antigen Symbol:FKBP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:O75344
- Antigen Synonyms:36 kDa FK506 binding protein|36 kDa FK506-binding protein|36 kDa FKBP|EC 5.2.1.8|FK506 binding protein 6|FK506-binding protein 6|FKBP 36|FKBP-36|FKBP-6|FKBP36|fkbp6|FKBP6_HUMAN|Immunophilin FKBP36|Peptidyl prolyl cis trans isomerase|Peptidyl-prolyl cis-trans isomerase FKBP6|PPIase|PPIase FKBP6|Rotamase
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Rotamase
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: FKBP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat