Anti-KPNB1 Rabbit Polyclonal Antibody
77741-830
New Product
- Antigen Symbol:KPNB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q14974
- Antigen Synonyms:IMB 1| IMB1| IMB1_HUMAN| Impnb| Importin 90| Importin beta 1| Importin beta 1 subunit| Importin subunit beta-1| Importin-90| IPOB| Karyopherin beta 1| Karyopherin beta 1 subunit| Karyopherin subunit beta-1| KPNB 1| MGC2155| MGC2156| MGC2157| NTF 97| NTF97| NTF97/Importin beta| Nuclear factor P97| Pore targeting complex 97 kDa subunit| PTAC97
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:PTAC97
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: KPNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse