Anti-HK2 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: HK2
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-098
- 77857-457
- Antigen Symbol:
- HK2
- HK2
- Antigen Synonyms:
- DKFZp686M1669| Hexokinase 2| Hexokinase 2 muscle| Hexokinase type II| Hexokinase-2| HK 2| HK II| HKII| HxK 2| HxK2| HXK2_HUMAN| Muscle form hexokinase
- DKFZp686M1669|Hexokinase 2|Hexokinase 2 muscle|Hexokinase type II|Hexokinase-2|HK 2|HK II|HKII|HxK 2|HxK2|HXK2_HUMAN|Muscle form hexokinase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52789
- P52789
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Hexokinase-2 protein (334-526AA)
- Recombinant Human Hexokinase-2 protein (334-526AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice