Anti-TBL2 Rabbit Polyclonal Antibody
Catalog # 77737-361
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:TBL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y4P3
- Antigen Synonyms:WBSCR13|UNQ563/PRO1125Transducin beta-like protein 2|WS beta-transducin repeats protein|WS-betaTRP|Williams-Beuren syndrome chromosomal region 13 protein
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Williams-Beuren syndrome chromosomal region 13 protein
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: TBL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human