Anti-FEM1A Rabbit Polyclonal Antibody
Catalog # 77737-040
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FEM1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9BSK4
- Antigen Synonyms:AW611390| DKFZp762M136| EPRAP| Fem-1 homolog a (C. elegans)| Fem1 homolog a| FEM1-alpha| FEM1-alpha-A| FEM1a-A| FEM1A_HUMAN| Fem1aa| Feminization 1 homolog a (C. elegans)| Feminization 1 homolog a| MGC114562| MGC117925| Prostaglandin E receptor 4-associated protein| Protein fem-1 homolog A| Protein fem-1 homolog A-A
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Protein fem-1 homolog A-A
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: FEM1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat