Anti-CCZ1/CCZ1B Rabbit Polyclonal Antibody
Catalog # 77736-674
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CCZ1/CCZ1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Monkey
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P86791/P86790
- Antigen Synonyms:CCZ1|C7orf28A|CGI-43|Vacuolar fusion protein CCZ1 homolog|CCZ1B|C7orf28B|Vacuolar fusion protein CCZ1 homolog B|Vacuolar fusion protein CCZ1 homolog-like
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:CCZ1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:10000
Type:
Antigen: CCZ1/CCZ1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Monkey