Anti-EML2 Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: EML2
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77735-854
- 77854-697
- Antigen Symbol:
- EML2
- EML2
- Antigen Synonyms:
- 1600029N02Rik| Echinoderm microtubule associated protein like 2| Echinoderm microtubule-associated protein-like 2| Echinoderm MT associated protein like protein 70| ELP70| EMAL2_HUMAN| EMAP-2| EMAP2| HuEMAP-2
- 1600029N02Rik|Echinoderm microtubule associated protein like 2|Echinoderm microtubule-associated protein-like 2|Echinoderm MT associated protein like protein 70|ELP70|EMAL2_HUMAN|EMAP-2|EMAP2|HuEMAP-2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95834
- O95834
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Echinoderm microtubule-associated protein-like 2 protein (1-418AA)
- Recombinant Human Echinoderm microtubule-associated protein-like 2 protein (1-418AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice